Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015436176.1 | XP_015291662.1 | 2 - 3 | 380 | 64 | |
XM_015436177.1 | XP_015291663.1 | 5 - 6 | 1293 | 64 | ||
XM_015436178.1 | XP_015291664.1 | 3 - 4 | 483 | 64 | ||
XM_015436179.1 | XP_015291665.1 | 2 - 3 | 391 | 64 | ||
XM_015436180.1 | XP_015291666.1 | 2 - 3 | 359 | 64 | ||
XM_015436181.1 | XP_015291667.1 | 2 - 3 | 368 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |