Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583069.2 | XP_005583126.1 | 3 - 4 | 1307 | 80 | |
XM_005583070.2 | XP_005583127.1 | 4 - 5 | 567 | 80 | ||
XM_005583071.2 | XP_005583128.1 | 4 - 5 | 569 | 80 | ||
XM_015437739.1 | XP_015293225.1 | 3 - 4 | 1277 | 80 | ||
XM_015437740.1 | XP_015293226.1 | 4 - 5 | 629 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |