Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGIR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589661.2 | XP_005589718.1 | 2 - 3 | 898 | 74 | |
XM_015441261.1 | XP_015296747.1 | 1 - 2 | 985 | 74 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Inventoried Probe spans exons |