Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590103.2 | XP_005590160.1 | 3 - 4 | 724 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |