Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H4orf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015450629.1 | XP_015306115.1 | 2 - 3 | 264 | 96 | |
XM_015450630.1 | XP_015306116.1 | 2 - 3 | 205 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |