Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H4orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556434.2 | XP_005556491.1 | 5 - 6 | 927 | 75 | |
XM_005556436.2 | XP_005556493.1 | 5 - 6 | 930 | 75 |