Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015454223.1 | XP_015309709.1 | 1 | 824 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |