Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005565891.2 | XP_005565948.1 | 9 - 10 | 1158 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |