Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVPR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319367.1 | NP_001306296.1 | 1 - 2 | 964 | 87 | |
GenBank mRNA | HM856594.1 | - | 1 - 2 | 964 | 87 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Probe spans exons |