Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMNTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015434204.1 | XP_015289690.1 | 7 - 8 | 1023 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |