Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005584693.2 | XP_005584750.1 | 2 - 3 | 148 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |