Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005542797.2 | XP_005542854.2 | 8 - 9 | 1536 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |