Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005559859.2 | XP_005559916.1 | 3 - 4 | 346 | 97 | |
XM_005559861.2 | XP_005559918.1 | 3 - 4 | 321 | 97 | ||
XM_005559862.2 | XP_005559919.1 | 3 - 4 | 477 | 97 | ||
XM_005559863.2 | XP_005559920.1 | 3 - 4 | 313 | 97 | ||
XM_005559864.2 | XP_005559921.1 | 3 - 4 | 331 | 97 | ||
XM_015452981.1 | XP_015308467.1 | 3 - 4 | 348 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |