Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005561768.2 | XP_005561825.1 | 1 - 2 | 139 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |