Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566868.2 | XP_005566925.1 | 1 | 1599 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |