Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567005.2 | XP_005567062.1 | 1 - 2 | 335 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |