Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BANF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568281.2 | XP_005568338.1 | 4 - 5 | 656 | 91 | |
XM_005568282.2 | XP_005568339.1 | 3 - 4 | 395 | 91 | ||
XM_015457773.1 | XP_015313259.1 | 4 - 5 | 782 | 91 | ||
XM_015457774.1 | XP_015313260.1 | 5 - 6 | 922 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |