Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571633.2 | XP_005571690.1 | 2 - 3 | 827 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |