Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588662.2 | XP_005588719.2 | 1 - 2 | 40 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |