Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015439049.1 | XP_015294535.1 | 23 - 24 | 2217 | 130 | |
GenBank mRNA | AB169152.1 | - | 13 - 14 | 1172 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Probe spans exons |