Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005560571.2 | XP_005560628.1 | 3 - 4 | 574 | 60 | |
XM_005560572.2 | XP_005560629.1 | 3 - 4 | 570 | 60 | ||
XM_005560573.1 | XP_005560630.1 | 3 - 4 | 323 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |