Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEBOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583220.2 | XP_005583277.2 | 2 - 3 | 358 | 88 | |
XM_015438044.1 | XP_015293530.1 | 2 - 3 | 319 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |