Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588092.2 | XP_005588149.2 | 2 - 3 | 226 | 130 | |
XM_005588093.2 | XP_005588150.1 | 4 - 5 | 439 | 130 | ||
XM_005588094.2 | XP_005588151.1 | 3 - 4 | 399 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |