Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284984.1 | NP_001271913.1 | 2 - 3 | 1101 | 81 | |
GenBank mRNA | AB056392.1 | - | 2 - 3 | 1101 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |