Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290711.1 | NP_001277640.1 | 8 - 9 | 978 | 60 | |
NM_007897.3 | NP_031923.1 | 8 - 9 | 859 | 60 | ||
GenBank mRNA | AK035781.1 | - | 8 - 9 | 742 | 60 | |
AK137389.1 | - | 8 - 9 | 900 | 60 | ||
BC139362.1 | - | 8 - 9 | 820 | 8862384 | 60 | |
BC139364.1 | - | 8 - 9 | 820 | 8862386 | 60 | |
BC139765.1 | - | 1 - 2 | 311 | 60 | ||
L12147.1 | - | 8 - 9 | 825 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |