Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007390.3 | NP_031416.3 | 2 - 3 | 248 | 110 | |
GenBank mRNA | AF225980.1 | - | 2 - 3 | 238 | 110 | |
AK034208.1 | - | 2 - 3 | 292 | 110 | ||
AK157809.1 | - | 2 - 3 | 245 | 110 | ||
AY574266.1 | - | 2 - 3 | 198 | 110 | ||
AY851289.1 | - | 73 | 110 | |||
L37663.1 | - | 2 - 3 | 248 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mus musculus Probe spans exons |