Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110204.1 | NP_001103674.1 | 6 - 7 | 996 | 100 | |
NM_001110205.1 | NP_001103675.1 | 4 - 5 | 474 | 100 | ||
NM_007394.3 | NP_031420.2 | 5 - 6 | 751 | 100 | ||
GenBank mRNA | AF332087.1 | - | 3 - 4 | 384 | 100 | |
AF332088.1 | - | 3 - 4 | 384 | 100 | ||
AK052246.1 | - | 586 | 100 | |||
AK053428.1 | - | 6 - 7 | 995 | 100 | ||
AK150014.1 | - | 5 - 6 | 751 | 100 | ||
AK150075.1 | - | 5 - 6 | 751 | 100 | ||
BC058718.1 | - | 3 - 4 | 472 | 6418954 | 100 | |
L15436.1 | - | 4 - 5 | 824 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |