Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198831.1 | NP_001185760.1 | 9 - 10 | 1275 | 60 | |
NM_001198833.1 | NP_001185762.1 | 9 - 10 | 1394 | 60 | ||
NM_007584.2 | NP_031610.1 | 9 - 10 | 1450 | 60 | ||
NM_172962.1 | NP_766550.1 | 9 - 10 | 1450 | 60 | ||
GenBank mRNA | AK031442.1 | - | 9 - 10 | 1353 | 60 | |
AK138698.1 | - | 9 - 10 | 1263 | 60 | ||
AK143088.1 | - | 9 - 10 | 1343 | 60 | ||
BC037108.1 | - | 10 - 11 | 1529 | 60 | ||
BC065998.1 | - | 9 - 10 | 1286 | 5686085 | 60 | |
L57509.1 | - | 9 - 10 | 1298 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |