Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007631.2 | NP_031657.1 | 1 - 2 | 426 | 73 | |
GenBank mRNA | AK005352.1 | - | 1 - 2 | 120 | 73 | |
AK017834.1 | - | 1 - 2 | 390 | 73 | ||
AK053348.1 | - | 1 - 2 | 334 | 73 | ||
AK149905.1 | - | 1 - 2 | 333 | 73 | ||
AK164997.1 | - | 1 - 2 | 371 | 73 | ||
AK170833.1 | - | 1 - 2 | 307 | 73 | ||
BC044841.1 | - | 1 - 2 | 329 | 3155470 | 73 | |
EU600170.1 | - | 1 - 2 | 426 | 73 | ||
M64403.1 | - | 1 - 2 | 381 | 73 | ||
S78355.1 | - | 1 - 2 | 331 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |