Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007854.3 | NP_031880.2 | 4 - 5 | 650 | 76 | |
GenBank mRNA | AF183397.1 | - | 4 - 5 | 560 | 76 | |
AK010950.1 | - | 4 - 5 | 587 | 76 | ||
AK083484.1 | - | 4 - 5 | 547 | 76 | ||
AK144451.1 | - | 4 - 5 | 543 | 76 | ||
AK154310.1 | - | 4 - 5 | 585 | 76 | ||
BC048958.1 | - | 4 - 5 | 796 | 6491511 | 76 | |
X86682.1 | - | 2 - 3 | 218 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |