Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007854.3 | NP_031880.2 | 8 - 9 | 1108 | 83 | |
GenBank mRNA | AF183397.1 | - | 8 - 9 | 1018 | 83 | |
AK010950.1 | - | 8 - 9 | 1045 | 83 | ||
AK083484.1 | - | 8 - 9 | 1005 | 83 | ||
AK144451.1 | - | 8 - 9 | 1001 | 83 | ||
AK154310.1 | - | 8 - 9 | 1043 | 83 | ||
BC048958.1 | - | 8 - 9 | 1254 | 6491511 | 83 | |
X86682.1 | - | 6 - 7 | 676 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |