Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dvl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007888.3 | NP_031914.3 | 10 - 11 | 1311 | 55 | |
GenBank mRNA | AK144001.1 | - | 10 - 11 | 1342 | 55 | |
AK146822.1 | - | 10 - 11 | 1311 | 55 | ||
AK159895.1 | - | 10 - 11 | 1144 | 55 | ||
AK168376.1 | - | 10 - 11 | 1219 | 55 | ||
AK219360.1 | - | 4 - 5 | 366 | 55 | ||
BC053050.1 | - | 10 - 11 | 1225 | 6402000 | 55 | |
BC092396.1 | - | 1318 | 30363483 | 55 | ||
U24160.1 | - | 10 - 11 | 1323 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |