Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007937.3 | NP_031963.2 | 1 - 2 | 1061 | 64 | |
GenBank mRNA | AK034289.1 | - | 1 - 2 | 1060 | 64 | |
AK049740.1 | - | 1 - 2 | 1062 | 64 | ||
AK089130.1 | - | 1 - 2 | 456 | 64 | ||
AK089143.1 | - | 1 - 2 | 920 | 64 | ||
BC057401.1 | - | 1 - 2 | 1039 | 5697758 | 64 | |
U07357.1 | - | 1 - 2 | 601 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |