Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007997.1 | NP_032023.1 | 6 - 7 | 697 | 69 | |
GenBank mRNA | AK134068.1 | - | 6 - 7 | 688 | 69 | |
BC057146.1 | - | 6 - 7 | 631 | 69 | ||
BC071268.1 | - | 6 - 7 | 697 | 6517167 | 69 | |
D49920.1 | - | 6 - 7 | 697 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |