Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163215.2 | NP_001156687.1 | 5 - 6 | 932 | 95 | |
NM_001163216.2 | NP_001156688.1 | 3 - 4 | 647 | 95 | ||
NM_001163217.2 | NP_001156689.1 | 4 - 5 | 701 | 95 | ||
NM_001205270.1 | NP_001192199.1 | 5 - 6 | 906 | 95 | ||
NM_008010.5 | NP_032036.2 | 5 - 6 | 906 | 95 | ||
GenBank mRNA | AF024638.1 | - | 3 - 4 | 555 | 95 | |
AK140217.1 | - | 4 - 5 | 823 | 95 | ||
AK143592.1 | - | 5 - 6 | 839 | 95 | ||
AK163495.1 | - | 3 - 4 | 485 | 95 | ||
AK163547.1 | - | 4 - 5 | 823 | 95 | ||
BC053056.1 | - | 5 - 6 | 816 | 5708838 | 95 | |
M81342.1 | - | 5 - 6 | 824 | 95 | ||
S56291.1 | - | 4 - 5 | 651 | 95 | ||
X58255.1 | - | 5 - 6 | 810 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |