Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008011.2 | NP_032037.2 | 8 - 9 | 1215 | 123 | |
GenBank mRNA | AK084850.1 | - | 8 - 9 | 1215 | 123 | |
AK085723.1 | - | 8 - 9 | 1213 | 123 | ||
AY493377.2 | - | 7 - 8 | 1101 | 123 | ||
BC033313.1 | - | 8 - 9 | 1402 | 5132831 | 123 | |
DQ388428.1 | - | 7 - 8 | 1101 | 123 | ||
X59927.1 | - | 8 - 9 | 1157 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |