Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162494.1 | NP_001155966.1 | 2 - 3 | 494 | 118 | |
NM_008056.3 | NP_032082.3 | 2 - 3 | 498 | 118 | ||
GenBank mRNA | AK088399.1 | - | 2 - 3 | 363 | 118 | |
BC026150.1 | - | 2 - 3 | 487 | 3983985 | 118 | |
CD348842.1 | - | 2 - 3 | 464 | 118 | ||
U43319.1 | - | 1 - 2 | 324 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |