Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162494.1 | NP_001155966.1 | 6 - 7 | 2283 | 87 | |
NM_008056.3 | NP_032082.3 | 6 - 7 | 2287 | 87 | ||
GenBank mRNA | AK088399.1 | - | 6 - 7 | 2152 | 87 | |
AK138964.1 | - | 3 - 4 | 1000 | 87 | ||
BC026150.1 | - | 6 - 7 | 2276 | 3983985 | 87 | |
U43319.1 | - | 5 - 6 | 2113 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Mus musculus Mouse Wnt Panel Inventoried Probe spans exons |