Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008067.4 | NP_032093.3 | 1 - 2 | 234 | 119 | |
GenBank mRNA | AK039144.1 | - | 1 - 2 | 175 | 119 | |
AK134760.1 | - | 1 - 2 | 143 | 119 | ||
M86568.1 | - | 1 - 2 | 136 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mus musculus 1700 Inventoried Probe spans exons |