Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008119.2 | NP_032145.2 | 2 - 3 | 191 | 67 | |
GenBank mRNA | AK008308.1 | - | 2 - 3 | 191 | 67 | |
U34295.1 | - | 1 - 2 | 94 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |