Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008168.2 | NP_032194.2 | 2 - 3 | 461 | 68 | |
GenBank mRNA | BC052009.1 | - | 2 - 3 | 349 | 5695215 | 68 |
BC058097.1 | - | 2 - 3 | 195 | 68 | ||
BC110682.1 | - | 2 - 3 | 384 | 5033584 | 68 | |
BU701817.1 | - | 2 - 3 | 461 | 68 | ||
CJ143126.1 | - | 2 - 3 | 437 | 68 | ||
D10011.1 | - | 2 - 3 | 390 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |