Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpind1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008223.3 | NP_032249.3 | 2 - 3 | 1049 | 121 | |
GenBank mRNA | BC034543.1 | - | 1 - 2 | 856 | 4158625 | 121 |
BC089610.1 | - | 2 - 3 | 1005 | 6768311 | 121 | |
CB949626.1 | - | 1 - 2 | 449 | 121 | ||
U07425.1 | - | 1 - 2 | 916 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |