Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110252.2 | NP_001103722.1 | 4 - 5 | 423 | 77 | |
NM_001276269.1 | NP_001263198.1 | 3 - 4 | 300 | 77 | ||
NM_008281.4 | NP_032307.2 | 3 - 4 | 363 | 77 | ||
GenBank mRNA | AF030065.1 | - | 3 - 4 | 322 | 77 | |
AK002694.1 | - | 4 - 5 | 381 | 77 | ||
AK156553.1 | - | 3 - 4 | 302 | 77 | ||
AY234104.1 | - | 3 - 4 | 322 | 77 | ||
BC138809.1 | - | 4 - 5 | 267 | 8861831 | 77 | |
BC145413.1 | - | 4 - 5 | 267 | 9053971 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |