Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008300.3 | NP_032326.3 | 11 - 12 | 1693 | 73 | |
GenBank mRNA | AK053601.1 | - | 11 - 12 | 1627 | 73 | |
AK054211.1 | - | 4 - 5 | 415 | 73 | ||
AK133772.1 | - | 11 - 12 | 1635 | 73 | ||
AK155305.1 | - | 11 - 12 | 1610 | 73 | ||
AK166102.1 | - | 11 - 12 | 1678 | 73 | ||
AK210383.1 | - | 3 - 4 | 336 | 73 | ||
AK220167.1 | - | 11 - 12 | 1644 | 73 | ||
BC003770.1 | - | 11 - 12 | 1602 | 3494648 | 73 | |
BC003950.1 | - | 11 - 12 | 1588 | 73 | ||
CN458707.1 | - | 5 - 6 | 635 | 73 | ||
D85904.1 | - | 11 - 12 | 1395 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |