Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196695 |
BC089571.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196695 |
NM_008372.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196695 |
AA183527.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196695 |
BC089571.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196695 |
NM_008372.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196695 |
AA183527.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
182425 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
178039 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
210595 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
716876 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
605109 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
62115 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
782010 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
896082 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
408292 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
380897 |
AK044445.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
380897 |
NM_172708.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
960221 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
633580 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
392087 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
424433 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
128703 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
157422 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
1090 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
171456 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P148026 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P100784 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196695 |
BC089571.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196695 |
NM_008372.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196695 |
AA183527.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196695 |
BC089571.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196695 |
NM_008372.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196695 |
AA183527.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P555957 |
M29697.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P555957 |
AK154516.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P555957 |
BC089571.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P555957 |
AK042264.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P555957 |
AK154215.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P32750 |
AK040740.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P32750 |
AK041838.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P196695 |
BC089571.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P196695 |
NM_008372.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P196695 |
AA183527.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P196695 |
BC089571.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P196695 |
NM_008372.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P196695 |
AA183527.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006040 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006055 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006022 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006027 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006006 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006015 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006045 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006025 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006049 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006009 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006038 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006051 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006037 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006041 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006043 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006021 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006054 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006017 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006050 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006023 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006019 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006020 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006034 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006014 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006018 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006035 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006011 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006046 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006007 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30006031 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08280544 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P148026 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08280504 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P100784 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P555957 |
M29697.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P555957 |
AK154516.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P555957 |
BC089571.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P555957 |
AK042264.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P555957 |
AK154215.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P32750 |
AK040740.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P32750 |
AK041838.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P196695 |
BC089571.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P196695 |
NM_008372.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P196695 |
AA183527.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P196695 |
BC089571.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P196695 |
NM_008372.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P196695 |
AA183527.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P104612 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P134958 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P148026 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P100784 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12336623 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12380887 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12336622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006035 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005843 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005937 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005845 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006050 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005859 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005876 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006046 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006022 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005893 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005980 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005930 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005863 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005917 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005994 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005850 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005986 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005836 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005919 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006064 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005869 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005884 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005842 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005924 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006003 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005975 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005959 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005887 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005886 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005943 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005922 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005958 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005861 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005992 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005910 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005905 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005972 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005966 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005894 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005918 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005879 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006043 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006004 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005881 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005891 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005984 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005912 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005926 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005964 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005898 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005896 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005946 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30006034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005955 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30005872 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006064 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12223780 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006046 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11975640 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006023 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006041 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005973 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005972 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006011 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12426611 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006006 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006019 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006007 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006040 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005922 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005992 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005987 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005976 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005970 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005975 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006050 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12010026 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006056 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006028 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006034 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12380891 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005928 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006031 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006003 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006054 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005986 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005927 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006004 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12400669 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006059 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006037 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005996 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005994 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005997 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005969 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006009 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005930 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006043 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11975638 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006038 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006035 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005932 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006025 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005971 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006051 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005984 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006049 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006017 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006018 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006066 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005982 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006020 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005977 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12020493 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30006021 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005920 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005980 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005924 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005990 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30005968 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12010024 |
- |
- |