Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008379.3 | NP_032405.3 | 16 - 17 | 2297 | 76 | |
GenBank mRNA | AK077709.1 | - | 16 - 17 | 2288 | 76 | |
AK147175.1 | - | 16 - 17 | 2316 | 76 | ||
AK169171.1 | - | 16 - 17 | 2264 | 76 | ||
BC052438.1 | - | 16 - 17 | 2052 | 6405935 | 76 | |
BC052711.1 | - | 16 - 17 | 2085 | 6835239 | 76 | |
BC055115.1 | - | 16 - 17 | 2282 | 6518750 | 76 | |
D45836.1 | - | 16 - 17 | 2188 | 76 | ||
D67015.1 | - | 16 - 17 | 2025 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |