Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008434.2 | NP_032460.2 | 10 - 11 | 1499 | 111 | |
GenBank mRNA | AB079603.1 | - | 10 - 11 | 1202 | 111 | |
AK136389.1 | - | 3 - 4 | 369 | 111 | ||
AK154244.1 | - | 10 - 11 | 1473 | 111 | ||
AY331142.1 | - | 10 - 11 | 1414 | 111 | ||
BC045142.1 | - | 10 - 11 | 1499 | 6314642 | 111 | |
BC055304.1 | - | 10 - 11 | 1454 | 6397978 | 111 | |
U70068.1 | - | 10 - 11 | 1305 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |