Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008465.5 | NP_032491.2 | 7 - 8 | 804 | 83 | |
GenBank mRNA | AK028259.1 | - | 7 - 8 | 805 | 83 | |
AK028307.1 | - | 7 - 8 | 823 | 83 | ||
AK154800.1 | - | 7 - 8 | 822 | 83 | ||
AK169311.1 | - | 7 - 8 | 827 | 83 | ||
AK169348.1 | - | 7 - 8 | 827 | 83 | ||
BC006771.1 | - | 7 - 8 | 789 | 3588534 | 83 | |
U20619.1 | - | 7 - 8 | 809 | 83 | ||
U34228.1 | - | 6 - 7 | 653 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |