Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400016953.mm.1 |
NM_008466.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016953.mm.1 |
NM_008466.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016953.mm.1 |
TC1400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016955.mm.1 |
NM_008466.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016955.mm.1 |
NM_008466.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016955.mm.1 |
TC1400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016958.mm.1 |
NM_008466.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016958.mm.1 |
NM_008466.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016958.mm.1 |
TC1400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016954.mm.1 |
NM_008466.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016954.mm.1 |
NM_008466.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016954.mm.1 |
TC1400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016956.mm.1 |
NM_008466.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016956.mm.1 |
NM_008466.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016956.mm.1 |
TC1400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016957.mm.1 |
NM_008466.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016957.mm.1 |
NM_008466.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016957.mm.1 |
TC1400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016959.mm.1 |
NM_008466.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016959.mm.1 |
NM_008466.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016959.mm.1 |
TC1400002189.mm.1 |
MTA TC |