Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008522.3 | NP_032548.2 | 9 - 10 | 1244 | 60 | |
GenBank mRNA | AK036491.1 | - | 9 - 10 | 1245 | 60 | |
AK144556.1 | - | 9 - 10 | 1245 | 60 | ||
AK150799.1 | - | 9 - 10 | 1245 | 60 | ||
AK151822.1 | - | 9 - 10 | 1245 | 60 | ||
AK164781.1 | - | 9 - 10 | 1246 | 60 | ||
BC006904.1 | - | 9 - 10 | 1252 | 3600033 | 60 | |
BC008530.1 | - | 9 - 10 | 1225 | 60 | ||
BC009662.1 | - | 5 - 6 | 675 | 60 | ||
CT010339.1 | - | 9 - 10 | 1210 | 60 | ||
D88510.1 | - | 9 - 10 | 1244 | 60 | ||
J03298.1 | - | 9 - 10 | 1213 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |